Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amer2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164705.1 | NP_001158177.1 | 2 | 1403 | ||
NM_028113.3 | NP_082389.1 | 1 | 1781 | |||
GenBank mRNA | AK011185.1 | - | 2 | 1338 | ||
AK032651.1 | - | 1 | 1722 | |||
AK139177.1 | - | 1 | 1728 | |||
BC030356.1 | - | 1 | 496 | |||
BC056350.1 | - | 2 | 1369 | 6415605 |