Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif1bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028197.2 | NP_082473.2 | 6 | 979 | ||
GenBank mRNA | AK035659.1 | - | 6 | 967 | ||
AK129320.1 | - | 6 | 964 | |||
AK141324.1 | - | 6 | 987 | |||
AK149629.1 | - | 6 | 916 | |||
AK158730.1 | - | 7 | 713 | |||
AK161007.1 | - | 965 | ||||
AK200964.1 | - | 18 | ||||
BC038828.1 | - | 2 | 288 | |||
BC046485.1 | - | 6 | 950 | 5350638 | ||
BC055307.1 | - | 6 | 933 | 6493547 | ||
BC090974.1 | - | 6 | 933 | 6774777 |