Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023672.2 | NP_076161.2 | 16, 17 | 1430 | ||
NM_198438.1 | NP_940840.1 | 15, 16 | 1349 | |||
GenBank mRNA | AF170906.1 | - | 15, 16 | 1079 | ||
AF500117.1 | - | 15, 16 | 1079 | |||
AK011509.1 | - | 16, 17 | 1045 | |||
AK011853.1 | - | 16, 17 | 1400 | |||
AK017711.1 | - | 16, 17 | 1434 | |||
AK041388.1 | - | 12, 13 | 946 | |||
AK133812.1 | - | 13, 14 | 888 | |||
AK153733.1 | - | 12, 13 | 790 | |||
AY167987.1 | - | 15, 16 | 962 | |||
BC003430.1 | - | 15, 16 | 1103 | 2646149 | ||
BC028324.1 | - | 10, 11 | 571 | |||
BC035475.1 | - | 15, 16 | 1292 |