Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175639.1 | NP_783570.1 | 2 | 304 | ||
GenBank mRNA | AK012043.1 | - | 2 | 289 | ||
AK032907.1 | - | 2 | 278 | |||
AK129031.1 | - | 2 | 222 | |||
AK152321.1 | - | 2 | 294 | |||
BY736005.1 | - | 2 | 304 |