Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shisa9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174086.1 | NP_001167557.1 | 1 | 775 | ||
NM_028277.2 | NP_082553.2 | 1 | 775 | |||
GenBank mRNA | AK012380.1 | - | 1 | 776 | ||
AK049860.1 | - | 1 | 301 | |||
BC030884.1 | - | 1 | 64 | |||
GU479981.1 | - | 1 | 307 | |||
GU479982.1 | - | 1 | 307 |