Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uaca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028283.2 | NP_082559.1 | 16 | 2534 | ||
GenBank mRNA | AB030647.1 | - | 16 | 2604 | ||
AK173203.1 | - | 4 | 2606 | |||
BC033470.1 | - | 17 | 1941 | 4951775 | ||
BC042415.1 | - | 16 | 2534 | 4165488 |