Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eldr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013290.1 | - | 4 | 744 | ||
BC037766.1 | - | 3 | 1332 | |||
BC054406.1 | - | 4 | 748 |