Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289671.1 | NP_001276600.1 | 1 | 290 | ||
NM_001289672.1 | NP_001276601.1 | 1 | 290 | |||
NM_001289673.1 | NP_001276602.1 | 1 | 290 | |||
GenBank mRNA | BC144818.1 | - | 1 | 290 | ||
BC144820.1 | - | 1 | 290 | |||
BC145799.1 | - | 1 | 290 | 40131102 |