Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PITRM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242307.1 | NP_001229236.1 | 15 | 1704 | ||
NM_001242309.1 | NP_001229238.1 | 14 | 1601 | |||
NM_014889.3 | NP_055704.2 | 15 | 1704 | |||
XM_005252345.3 | XP_005252402.1 | 15 | 1704 | |||
XM_017015470.1 | XP_016870959.1 | 15 | 1772 | |||
XM_017015471.1 | XP_016870960.1 | 15 | 1772 | |||
XM_017015472.1 | XP_016870961.1 | 15 | 1601 | |||
XM_017015473.1 | XP_016870962.1 | 15 | 1601 | |||
GenBank mRNA | AB029027.2 | - | 5 | 433 | ||
AF061243.1 | - | 15 | 1648 | |||
AK002061.1 | - | 15 | 1672 | |||
AK023476.1 | - | 8 | 819 | |||
AK225084.1 | - | 15 | 1642 | |||
AK294865.1 | - | 14 | 1617 | |||
AK299464.1 | - | 14 | 1450 | |||
AK303406.1 | - | 14 | 1542 | |||
AK315117.1 | - | 15 | 1642 | |||
BC001150.1 | - | 9 | 1004 | 3503609 | ||
BC005025.1 | - | 15 | 1639 | 4040225 | ||
BC045577.1 | - | 15 | 2114 | |||
BC095422.1 | - | 15 | 1645 | 4822631 | ||
BC111987.1 | - | 15 | 1660 | 8327455 | ||
BC113369.1 | - | 15 | 1660 | 8322421 | ||
BX649017.1 | - | 15 | 1646 | |||
CR749279.1 | - | 13 | 1380 |