Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081435.1 | NP_001074904.1 | 10 | 1468 | ||
GenBank mRNA | AK013672.1 | - | 1 | 170 | ||
BC099560.1 | - | 1 | 164 | |||
BC158049.1 | - | 10 | 1403 | 9007353 |