Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clhc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163521.1 | NP_001156993.1 | 5 | 817 | ||
NM_028504.1 | NP_082780.1 | 5 | 850 | |||
GenBank mRNA | AK006591.1 | - | 5 | 851 | ||
BC137683.1 | - | 5 | 774 | 8860702 | ||
BC144828.1 | - | 5 | 741 | 9053376 |