Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tubb2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023716.2 | NP_076205.1 | 2 | 245 | ||
GenBank mRNA | AK010786.1 | - | 2 | 246 | ||
AK168908.1 | - | 2 | 251 | |||
BC138935.1 | - | 2 | 232 | 8861957 | ||
BC138936.1 | - | 2 | 232 | 8861958 |
Gene Symbol: Tubb2a-ps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BG066514.2 | - | 1 | 359 |