Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNASEH2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006397.2 | NP_006388.2 | 4, 5 | 479 | ||
XM_006722619.2 | XP_006722682.1 | 5, 6 | 664 | |||
GenBank mRNA | AK315327.1 | - | 4, 5 | 509 | ||
AY363912.1 | - | 4, 5 | 468 | |||
BC011748.2 | - | 4, 5 | 470 | 3160621 | ||
BG825340.1 | - | 4, 5 | 483 | |||
CR456841.1 | - | 4, 5 | 419 | |||
Z97029.1 | - | 4, 5 | 479 |