Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cluh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081158.2 | NP_001074627.1 | 15 | 2693 | ||
GenBank mRNA | AB093256.1 | - | 13 | 2240 | ||
AK004841.1 | - | 3 | 241 | |||
AK036540.1 | - | 15 | 2677 | |||
AK145708.1 | - | 14 | 2582 | |||
AK150045.1 | - | 15 | 2653 | |||
AK154146.1 | - | 15 | 2682 | |||
AK154925.1 | - | 15 | 2694 | |||
AK157310.1 | - | 15 | 2703 | |||
AK207283.1 | - | 1 | 14 | |||
BC060069.1 | - | 10 | 1805 | |||
BC072573.1 | - | 14 | 3290 | 5705116 | ||
BC094446.1 | - | 6 | 1273 |