Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ranbp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145824.4 | NP_665823.2 | 1 | 390 | ||
GenBank mRNA | AK048772.1 | - | 1 | 246 | ||
AK173177.1 | - | 1 | 203 | |||
AY337314.1 | - | 1 | 391 | |||
BC024698.1 | - | 1 | 235 | 3667000 | ||
EU281316.1 | - | 1 | 251 |