Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Neto2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081324.1 | NP_001074793.1 | 9 | 1635 | ||
GenBank mRNA | AK017458.1 | - | 9 | 1712 | ||
AK081462.1 | - | 9 | 1734 | |||
AK083512.1 | - | 9 | 1765 | |||
AY138991.1 | - | 9 | 1599 | |||
BC068168.1 | - | 10 | 1811 | |||
BC151050.1 | - | 9 | 1595 | 9087963 | ||
BC157941.1 | - | 9 | 1574 | 9088121 | ||
BC172104.1 | - | 9 | 1574 | 9054390 | ||
BC172139.1 | - | 9 | 1574 | 9054809 |