Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Manf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029103.3 | NP_083379.2 | 3 | 383 | ||
GenBank mRNA | AB009291.1 | - | 3 | 357 | ||
AK014338.1 | - | 3 | 361 | |||
AK131997.1 | - | 3 | 384 | |||
AK165646.1 | - | 3 | 359 | |||
AK167580.1 | - | 3 | 383 | |||
AK176996.1 | - | 3 | 277 | |||
AK186239.1 | - | 3 | 278 | |||
BC022761.1 | - | 3 | 389 | |||
BC038901.1 | - | 3 | 371 | |||
BC048616.1 | - | 2 | 446 | 6742574 |