Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uhrf1bp1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029166.2 | NP_083442.2 | 10 | 1455 | ||
GenBank mRNA | AB093261.1 | - | 10 | 1420 | ||
AK087280.1 | - | 8 | 1007 | |||
AK141050.1 | - | 10 | 1464 | |||
BC030383.1 | - | 1414 | ||||
BC079645.1 | - | 3 | 266 | |||
BC132134.1 | - | 10 | 1248 | 40130411 | ||
BC138092.1 | - | 10 | 1248 | 8861111 |