Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asxl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270988.1 | NP_001257917.1 | 8 | 1143 | ||
NM_172421.5 | NP_766009.2 | 7 | 999 | |||
GenBank mRNA | AK016141.1 | - | 7 | 705 | ||
AK035679.1 | - | 2 | 1571 | |||
AK036839.1 | - | 7 | 956 | |||
AK080679.1 | - | 8 | 1117 | |||
AK148015.1 | - | 7 | 1004 | |||
BC141140.1 | - | 8 | 977 | 9055752 |