Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029280.4 | NP_083556.2 | 1 | 309 | ||
GenBank mRNA | AA466649.1 | - | 1 | 307 | ||
AK013063.1 | - | 1 | 292 | |||
AK133594.1 | - | 1 | 299 | |||
BC027547.1 | - | 1 | 242 | 1531973 | ||
CJ063020.1 | - | 1 | 299 |