Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Faxc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175234.4 | NP_780443.2 | 3 | 687 | ||
GenBank mRNA | AK018081.1 | - | 3 | 656 | ||
AK082464.1 | - | 3 | 681 | |||
AK144924.1 | - | 3 | 688 | |||
BC059864.1 | - | 3 | 201 |