Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029617.2 | NP_083893.2 | 8 | 1428 | ||
GenBank mRNA | AK009772.1 | - | 8 | 1430 | ||
AK033132.1 | - | 8 | 1732 | |||
BC080815.1 | - | 8 | 1746 | 30104270 |