Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arfrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165991.1 | NP_001159463.1 | 4 | 436 | ||
NM_001165992.1 | NP_001159464.1 | 4 | 417 | |||
NM_001165995.1 | NP_001159467.1 | 3 | 323 | |||
NM_029702.4 | NP_083978.3 | 4 | 430 | |||
GenBank mRNA | AJ413953.1 | - | 4 | 383 | ||
AK005174.1 | - | 4 | 378 | |||
AK044759.1 | - | 4 | 388 | |||
AK050136.1 | - | 1 | 1288 | |||
AK083490.1 | - | 1 | 273 | |||
AK185500.1 | - | 1 | 36 | |||
BC010713.1 | - | 4 | 377 | 2649462 | ||
BC046782.1 | - | 4 | 368 | 5720540 | ||
BC046786.1 | - | 4 | 368 | 6416321 | ||
CA539216.1 | - | 4 | 430 |