Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtif2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282118.1 | NP_001269047.1 | 7 | 1029 | ||
NM_001282119.1 | NP_001269048.1 | 7 | 951 | |||
NM_001282120.1 | NP_001269049.1 | 7 | 1393 | |||
NM_133767.3 | NP_598528.2 | 7 | 1307 | |||
GenBank mRNA | AK010766.1 | - | 7 | 1252 | ||
AK136344.1 | - | 7 | 991 | |||
AK146961.1 | - | 7 | 1247 | |||
BC016590.1 | - | 7 | 883 | 3585359 | ||
BC066819.1 | - | 7 | 1287 |