Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMTOR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006402.2 | NP_006393.2 | 1 | 176 | ||
GenBank mRNA | BC062619.1 | - | 1 | 194 | 5190406 | |
BX092018.1 | - | 1 | 192 |