Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spopl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165997.1 | NP_001159469.1 | 12 | 1464 | ||
NM_001165998.1 | NP_001159470.1 | 11 | 1328 | |||
NM_029773.2 | NP_084049.2 | 11 | 1367 | |||
GenBank mRNA | AK014913.1 | - | 6 | 425 | ||
AK162056.1 | - | 11 | 1182 | |||
BC111867.1 | - | 12 | 1338 | 40056321 | ||
CD541905.1 | - | 6 | 461 |