Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spopl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165997.1 | NP_001159469.1 | 8, 9 | 1139 | ||
NM_001165998.1 | NP_001159470.1 | 7, 8 | 1003 | |||
NM_029773.2 | NP_084049.2 | 7, 8 | 1042 | |||
GenBank mRNA | AK014913.1 | - | 2, 3 | 100 | ||
AK162056.1 | - | 7, 8 | 857 | |||
BC111867.1 | - | 8, 9 | 1013 | 40056321 | ||
CD541905.1 | - | 2, 3 | 136 | |||
CF746115.1 | - | 4, 5 | 496 |