Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufaf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085493.2 | NP_001078962.1 | 1 | 75 | ||
GenBank mRNA | AK009547.1 | - | 1 | 55 | ||
AK041507.1 | - | 1 | 58 |