Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jrkl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033181.1 | NP_001028353.1 | 1 | 795 | ||
GenBank mRNA | BC138472.1 | - | 1 | 795 | 8861492 | |
BC138475.1 | - | 1 | 795 | 8861495 |