Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snapc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029949.3 | NP_084225.1 | 7 | 861 | ||
GenBank mRNA | AK007563.1 | - | 7 | 861 | ||
AK019863.2 | - | 7 | 857 | |||
AK031870.1 | - | 7 | 863 | |||
AK031924.1 | - | 6 | 779 | |||
AK049933.1 | - | 7 | 882 | |||
BC048713.1 | - | 7 | 861 | 6330194 |