Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krtap19-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130857.2 | NP_570927.1 | 1 | 330 | ||
GenBank mRNA | AF345295.1 | - | 1 | 282 | ||
AK020697.1 | - | 1 | 331 | |||
BC115545.2 | - | 1 | 282 | 40101652 | ||
BC115546.2 | - | 1 | 282 | 40101653 |