Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRDX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006406.1 | NP_006397.1 | 3 | 457 | ||
XM_005274438.1 | XP_005274495.1 | 3 | 632 | |||
XM_017029230.1 | XP_016884719.1 | 3 | 434 | |||
XM_017029231.1 | XP_016884720.1 | 3 | 632 | |||
GenBank mRNA | BC003609.2 | - | 3 | 417 | 3508414 | |
BC007107.1 | - | 3 | 461 | 4284320 | ||
BC016770.1 | - | 3 | 471 | 4075326 | ||
CR541668.1 | - | 3 | 414 | |||
CR541705.1 | - | 3 | 414 | |||
FJ224297.1 | - | 3 | 457 | |||
U25182.1 | - | 3 | 457 |