Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL6IP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006407.3 | NP_006398.1 | 3 | 569 | ||
GenBank mRNA | AB014765.1 | - | 3 | 475 | ||
AB052638.1 | - | 3 | 467 | |||
AB097051.1 | - | 3 | 550 | |||
AF070523.1 | - | 3 | 575 | |||
AF115965.1 | - | 3 | 560 | |||
AF125530.1 | - | 3 | 561 | |||
AF161476.1 | - | 3 | 550 | |||
AK074612.1 | - | 3 | 521 | |||
AK223566.1 | - | 3 | 473 | |||
AK301637.1 | - | 4 | 483 | |||
AK303148.1 | - | 3 | 455 | |||
AK312731.1 | - | 3 | 472 | |||
AY102608.1 | - | 3 | 460 | |||
BC005143.2 | - | 3 | 492 | 3510436 | ||
BC020797.1 | - | 3 | 486 | 4134658 |