Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tusc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019742.4 | NP_062716.1 | 2, 3 | 406 | ||
GenBank mRNA | AF123387.1 | - | 2, 3 | 293 | ||
AK004430.1 | - | 2, 3 | 366 | |||
AK082684.1 | - | 2, 3 | 407 | |||
BC048477.1 | - | 2, 3 | 406 | 6772171 |