Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290688.1 | NP_001277617.1 | 2 | 144 | ||
NM_020000.3 | NP_064384.2 | 2 | 144 | |||
NM_173719.3 | NP_776067.1 | 2 | 144 | |||
GenBank mRNA | AB041805.1 | - | 1 | 1600 | ||
AK005422.1 | - | 2 | 127 | |||
AK008759.1 | - | 2 | 123 | |||
AK011080.1 | - | 2 | 144 | |||
AK147032.1 | - | 2 | 130 | |||
AK148025.1 | - | 2 | 125 | |||
BC002315.1 | - | 1 | 84 | |||
BC021870.1 | - | 2 | 115 | 4022630 | ||
BC051683.1 | - | 2 | 122 | |||
BC053738.1 | - | 1 | 337 |