Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dtx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030714.2 | NP_109639.1 | 5 | 1223 | ||
GenBank mRNA | AB015425.1 | - | 4 | 1205 | ||
AK148444.1 | - | 5 | 1242 | |||
AK180250.1 | - | 1 | 23 | |||
AK180771.1 | - | 1 | 23 | |||
AK181396.1 | - | 1 | 23 | |||
AK183648.1 | - | 1 | 23 | |||
AK189235.1 | - | 1 | 23 | |||
AK195108.1 | - | 1 | 23 | |||
AK205640.1 | - | 1 | 41 | |||
AK207039.1 | - | 1 | 23 | |||
AK218545.1 | - | 1 | 23 | |||
AK220046.1 | - | 1 | 23 | |||
BC099687.1 | - | 5 | 1223 | 6311735 | ||
BC138304.1 | - | 4 | 1102 | 8861323 | ||
BC157899.1 | - | 4 | 1104 | 9088091 |