Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pum2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160219.1 | NP_001153691.1 | 19 | 3038 | ||
NM_001160220.1 | NP_001153692.1 | 19 | 3032 | |||
NM_001160221.1 | NP_001153693.1 | 19 | 2847 | |||
NM_001160222.1 | NP_001153694.1 | 18 | 2610 | |||
NM_030723.2 | NP_109648.2 | 19 | 2962 | |||
GenBank mRNA | AF315590.1 | - | 19 | 2978 | ||
AK048119.1 | - | 13 | 1901 | |||
AK122225.1 | - | 20 | 2880 | |||
AK134185.1 | - | 19 | 2846 | |||
AK141682.1 | - | 19 | 2894 | |||
AK147617.1 | - | 20 | 2980 | |||
AK161357.1 | - | 8 | 1203 | |||
AK163964.1 | - | 18 | 2585 | |||
AY027917.1 | - | 19 | 2878 | |||
BC013765.1 | - | 3 | 213 | |||
BC041773.1 | - | 17 | 2584 | 3986207 |