Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pum2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160219.1 | NP_001153691.1 | 15 | 2575 | ||
NM_001160220.1 | NP_001153692.1 | 15 | 2575 | |||
NM_001160221.1 | NP_001153693.1 | 15 | 2390 | |||
NM_001160222.1 | NP_001153694.1 | 14 | 2153 | |||
NM_030723.2 | NP_109648.2 | 15 | 2499 | |||
GenBank mRNA | AF315590.1 | - | 15 | 2515 | ||
AK048119.1 | - | 9 | 1444 | |||
AK122225.1 | - | 16 | 2423 | |||
AK134185.1 | - | 15 | 2389 | |||
AK141682.1 | - | 15 | 2437 | |||
AK147617.1 | - | 16 | 2523 | |||
AK161357.1 | - | 4 | 746 | |||
AK163964.1 | - | 14 | 2128 | |||
AY027917.1 | - | 15 | 2415 | |||
BC041773.1 | - | 13 | 2127 | 3986207 |