Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jam3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023277.4 | NP_075766.1 | 2 | 185 | ||
GenBank mRNA | AJ300304.1 | - | 2 | 145 | ||
AK003326.1 | - | 2 | 145 | |||
AK013156.1 | - | 2 | 144 | |||
AK032833.1 | - | 2 | 145 | |||
AK160648.1 | - | 2 | 183 | |||
AW106511.1 | - | 2 | 185 | |||
BC024357.1 | - | 2 | 157 | 4973185 |