Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006408.3 | NP_006399.1 | 4, 5 | 444 | ||
XM_005249581.4 | XP_005249638.1 | 4, 5 | 413 | |||
GenBank mRNA | AF007791.1 | - | 3, 4 | 267 | ||
AF038451.1 | - | 4, 5 | 315 | |||
AF087879.1 | - | 4, 5 | 406 | |||
AF088867.1 | - | 4, 5 | 404 | |||
AF115926.1 | - | 4, 5 | 314 | |||
AK223067.1 | - | 4, 5 | 335 | |||
AK312942.1 | - | 4, 5 | 325 | |||
AY359009.1 | - | 4, 5 | 404 | |||
BC015503.1 | - | 4, 5 | 313 | 3852448 | ||
BT007048.1 | - | 3, 4 | 257 | |||
FJ224307.1 | - | 3, 4 | 267 | |||
KJ767789.1 | - | 4, 5 | 418 |