Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpv17l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289561.1 | NP_001276490.1 | 4 | 1124 | ||
NM_001289562.1 | NP_001276491.1 | 4 | 856 | |||
NM_001289563.1 | NP_001276492.1 | 4 | 819 | |||
NM_001289565.1 | NP_001276494.1 | 5 | 873 | |||
NM_001289567.1 | NP_001276496.1 | 5 | 1149 | |||
NM_033564.3 | NP_291042.2 | 4 | 1095 | |||
GenBank mRNA | AF305634.1 | - | 4 | 1065 | ||
AK034151.1 | - | 1 | 571 | |||
AK136449.1 | - | 4 | 857 | |||
AK144372.1 | - | 4 | 818 | |||
AY513273.1 | - | 4 | 983 | |||
BC037713.1 | - | 4 | 975 | |||
BC094450.1 | - | 4 | 1062 | 4973897 |