Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053159.3 | NP_444389.2 | 8 | 936 | ||
GenBank mRNA | AB049633.1 | - | 8 | 817 | ||
AK002509.1 | - | 8 | 906 | |||
AK008592.1 | - | 8 | 966 | |||
AK054185.1 | - | 5 | 371 | |||
AK077857.1 | - | 8 | 936 | |||
AK145687.1 | - | 8 | 966 | |||
AK169272.1 | - | 8 | 937 | |||
BC011128.1 | - | 8 | 905 | 4187019 | ||
BC016586.1 | - | 8 | 809 | 2616397 | ||
BC034877.1 | - | 8 | 902 | 1547459 | ||
KC508086.1 | - | 8 | 913 |