Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053164.3 | NP_444394.1 | 3 | 766 | ||
GenBank mRNA | AA253916.1 | - | 1 | 28 | ||
AB049657.1 | - | 3 | 726 | |||
AK015357.1 | - | 3 | 747 | |||
AK076204.1 | - | 3 | 725 | |||
BC027676.1 | - | 3 | 706 | |||
BC048733.1 | - | 3 | 757 | 6512868 | ||
BC065812.1 | - | 3 | 713 |