Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4933434E20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287086.1 | NP_001274015.1 | 4 | 610 | ||
NM_001287087.1 | NP_001274016.1 | 8 | 927 | |||
NM_025762.3 | NP_080038.1 | 6 | 782 | |||
NM_027500.4 | NP_081776.2 | 6 | 782 | |||
GenBank mRNA | AK017833.1 | - | 6 | 750 | ||
AK034749.1 | - | 6 | 745 | |||
AK051238.1 | - | 6 | 675 | |||
AK077215.1 | - | 8 | 928 | |||
AK150324.1 | - | 6 | 733 | |||
AK150423.1 | - | 6 | 733 | |||
AK150831.1 | - | 6 | 733 | |||
AK151002.1 | - | 6 | 732 | |||
AK151263.1 | - | 6 | 731 | |||
AY533135.1 | - | 6 | 530 | |||
BC018438.1 | - | 4 | 540 | 2609299 | ||
BC022598.1 | - | 1 | 15 | |||
BC027190.1 | - | 6 | 642 | 3257131 | ||
BG091444.1 | - | 4 | 377 | |||
BM935139.1 | - | 1 | 225 |