Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168398.1 | NP_001161870.1 | 2 | 156 | ||
NM_006416.4 | NP_006407.1 | 2 | 156 | |||
GenBank mRNA | AJ810302.1 | - | 2 | 92 | ||
AJ810303.1 | - | 2 | 92 | |||
AJ851888.1 | - | 2 | 77 | |||
AJ851889.1 | - | 2 | 77 | |||
AJ851890.1 | - | 2 | 77 | |||
AK293696.1 | - | 2 | 108 | |||
AK299432.1 | - | 2 | 104 | |||
AL122119.1 | - | 2 | 152 | |||
BC017807.1 | - | 2 | 104 | 4690513 | ||
BG699364.1 | - | 2 | 161 | |||
D87969.1 | - | 2 | 104 | |||
DA210713.1 | - | 2 | 162 | |||
DB095595.1 | - | 2 | 156 |