Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006416.4 | NP_006407.1 | 5, 6 | 652 | ||
GenBank mRNA | AJ810302.1 | - | 5, 6 | 586 | ||
AJ810303.1 | - | 5, 6 | 588 | |||
AJ851890.1 | - | 4, 5 | 413 | |||
AK293696.1 | - | 4, 5 | 444 | |||
BC017807.1 | - | 5, 6 | 600 | 4690513 | ||
D87969.1 | - | 5, 6 | 600 |