Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009570.2 | NP_001009570.1 | 3 | 355 | ||
NM_001166284.1 | NP_001159756.1 | 6 | 706 | |||
NM_001166285.1 | NP_001159757.1 | 9 | 1063 | |||
NM_006429.3 | NP_006420.1 | 8 | 967 | |||
NR_029402.1 | 7 | 914 | ||||
NR_029403.1 | 7 | 818 | ||||
XM_011532478.2 | XP_011530780.1 | 9 | 997 | |||
XM_011532479.1 | XP_011530781.1 | 8 | 900 | |||
GenBank mRNA | AF026292.1 | - | 8 | 892 | ||
AK098549.1 | - | 8 | 885 | |||
AK098825.1 | - | 3 | 299 | |||
AK222478.1 | - | 8 | 875 | |||
AK291961.1 | - | 8 | 913 | |||
AK293175.1 | - | 7 | 832 | |||
AK293597.1 | - | 3 | 283 | |||
AK297846.1 | - | 9 | 991 | |||
AK298153.1 | - | 6 | 634 | |||
AK302078.1 | - | 7 | 746 | |||
BC019296.1 | - | 8 | 880 | 2905446 | ||
BC088351.1 | - | 8 | 922 | 6202511 | ||
BM558355.1 | - | 255 | ||||
CR456719.1 | - | 7 | 824 | |||
CR536511.1 | - | 7 | 824 | |||
U83843.1 | - | 7 | 788 |