Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNLY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302758.1 | NP_001289687.1 | 4 | 417 | ||
NM_006433.4 | NP_006424.2 | 3 | 336 | |||
NM_012483.3 | NP_036615.2 | 4 | 580 | |||
XM_005264084.2 | XP_005264141.1 | 4 | 350 | |||
XM_005264085.2 | XP_005264142.1 | 3 | 325 | |||
XM_005264087.2 | XP_005264144.1 | 4 | 577 | |||
GenBank mRNA | AK304728.1 | - | 4 | 350 | ||
AK308781.1 | - | 3 | 439 | |||
AK310057.1 | - | 4 | 578 | |||
AK311012.1 | - | 3 | 289 | |||
AK311537.1 | - | 4 | 470 | |||
AK311538.1 | - | 3 | 203 | |||
AK311657.1 | - | 3 | 866 | |||
BC023576.2 | - | 3 | 211 | 4766109 | ||
CR541859.1 | - | 3 | 208 | |||
X05044.1 | - | 4 | 440 | |||
X54101.1 | - | 3 | 336 |