Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAB21L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006439.4 | NP_006430.1 | 1 | 1556 | ||
GenBank mRNA | AF262032.1 | - | 1 | 1206 | ||
AF370007.1 | - | 1 | 457 | |||
AK055665.1 | - | 1 | 1330 | |||
BC009983.2 | - | 1 | 1054 | 4125321 |