Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTHFS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199758.1 | NP_001186687.1 | 3 | 423 | ||
NM_006441.3 | NP_006432.1 | 3 | 476 | |||
NR_037654.1 | 3 | 505 | ||||
GenBank mRNA | AK054972.1 | - | 1 | 1781 | ||
AL110101.1 | - | 3 | 422 | |||
BC012417.1 | - | 3 | 386 | |||
BC019921.1 | - | 3 | 476 | 3900731 | ||
BM450865.1 | - | 3 | 493 | |||
CB995322.1 | - | 3 | 449 | |||
L38928.1 | - | 3 | 428 |
Gene Symbol: ST20-MTHFS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199760.1 | NP_001186689.1 | 4 | 422 | ||
GenBank mRNA | BG705061.1 | - | 3 | 602 | ||
CK002935.1 | - | 4 | 426 | |||
DB495490.1 | - | 2 | 337 |